Order Entry
Puerto Rico
ContactUsLinkComponent
Quantofix Chlorine Sensitive Test Strips, CTL Scientific Supply
Quantofix Chlorine Sensitive Test Strips, CTL Scientific Supply
Catalog # 10217-542
Supplier:  Macherey-Nagel
CAS Number:  
undefined
Quantofix Chlorine Sensitive Test Strips, CTL Scientific Supply
Catalog # 10217-542
Supplier:  Macherey-Nagel
Supplier Number:  91339
CAS Number:  

Specifications

  • Description:
    Chlorine Sensitive Test Strips
  • Storage:
    RT
  • Cat. No.:
    10217-542
  • Range:
    0-0.1-0.5-1-3-10 mg/L
  • Supplier no.:
    91339

Specifications

About this item

Quantofix Chlorine Sensitive test strips for the semi-quantitative determination of total chlorine.

Quantofix Chlorine Sensitive has a gradation of scale: 0-0.1-0.5-1-3-10 mg/L

Ordering information: Packaged as a box of 100.