Order Entry
Puerto Rico
ContactUsLinkComponent
Quantofix Molybdenum Test Strips, CTL Scientific Supply
Quantofix Molybdenum Test Strips, CTL Scientific Supply
Catalog # 10217-524
Supplier:  Macherey-Nagel
CAS Number:  
undefined
Quantofix Molybdenum Test Strips, CTL Scientific Supply
Catalog # 10217-524
Supplier:  Macherey-Nagel
Supplier Number:  91325
CAS Number:  

Specifications

  • Description:
    Molybdenum Test Strips
  • Storage:
    RT
  • Cat. No.:
    10217-524
  • Range:
    0-5-20-50-100-250 mg/L
  • Supplier no.:
    91325

Specifications

About this item

Quantofix Molybdenum test strips and reagents for the semi-quantitative determination of molybdenum.

Quantofix Molybdenum has a gradation of scale: 0-5-20-50-100-250 mg/L Mo6+ white to green. Store molybdenum test sticks in a cool and dry place, protected from sunlight and moisture (storage temperature below + 30 °C).

Ordering information: Packaged as a box of 100.