Quantofix Cyanide Test Kit, CTL Scientific Supply
Catalog # 10217-514
Supplier: Macherey-Nagel
CAS Number:
Specifications
- Description:Cyanide Test Strips and Reagents
- Storage:RT
- Cat. No.:10217-514
- Range:0-1-3-10-30 mg/L
- Supplier no.:91318
Specifications
About this item
Quantofix Cyanide test strips and reagents for the semi-quantitative determination of cyanide in solutions.
Quantofix Cyanide has a gradation of scale: 0-1-3-10-30 mg/L.
Ordering information: Packaged as a box of 100, strips and reagent.