Order Entry
Puerto Rico
ContactUsLinkComponent
Quantofix Cobalt Test Strips, CTL Scientific Supply
Quantofix Cobalt Test Strips, CTL Scientific Supply
Catalog # 10217-510
Supplier:  Macherey-Nagel
CAS Number:  
undefined
Quantofix Cobalt Test Strips, CTL Scientific Supply
Catalog # 10217-510
Supplier:  Macherey-Nagel
Supplier Number:  91303
CAS Number:  

Specifications

  • Description:
    Cobalt Test Strips
  • Storage:
    RT
  • Cat. No.:
    10217-510
  • Range:
    0-10-25-50-100-250-500-1000 mg/L
  • Supplier no.:
    91303

Specifications

About this item

Quantofix Cobalt Test strips for the semi-quantitative determination of cobalt in solutions.

Quantofix Cobalt Test strips has a gradation of scale: 0-10-25-50-100-250-500-1000 mg/L.

Ordering information: Packaged as a box of 100.