CENCO® Drilled Ball Set
Catalog # 470164-714
Supplier: United Scientific Supplies
Specifications
- Description:CENCO® Drilled Ball Set
- Cat. No.:470164-714
- Supplier no.:DBLST12
Specifications
About this item
High Quality.
- Every set comes in a convenient plastic storage case, with a foam bottom that holds each ball firmly in place
Each set contains 2 each of brass, aluminum, steel, wood, cork, and copper balls. A 3mm hole is drilled through the center of each ball. A slightly larger hole is drilled half way through the center of the ball. This allows the knot of a string to be held at the center of mass of the ball for superior peformance in oscillation experiments.