ContactUsLinkComponent
Stainless Steel Specimen Chuck Stand, Springside Scientific
Stainless Steel Specimen Chuck Stand, Springside Scientific
Catalog # 10764-046
undefined
Stainless Steel Specimen Chuck Stand, Springside Scientific
Catalog # 10764-046
Supplier Number:  SDST001
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Specimen Chuck Stand
  • Dimensions:
    8 7/8 x 2 3/4 x 1 1/4"
  • Cat. No.:
    10764-046
  • Supplier no.:
    SDST001
Specifications
About this item
This stainless steel stand is used to store chucks both outside and inside the cooling chamber.
This small metal stand fits five specimen chucks up to 55 mm in diameter. Specimen chucks are not included. High quality stainless steel stand is perfect for in-chamber and can be used to expand the internal chuck capacity of the cryostat.