GeneMate Individual PCR Tubes, with Frosted Flat Caps, 0.5 ml, Scientific Specialties
Catalog # 490004-478
Supplier: GeneMate
Specifications
- Capacity:0.5 ml
- Color:Natural
- Description:PCR tubes, with frosted flat caps
- Cat. No.:490004-478
- Supplier no.:3320-00-110IS
Specifications
About this item
Secure evaporation-resistant seal between cap lid and tube.
- Ultra-thin wall for optimum heat transfer and higher yield
- Crystal clear clarity allows easy content viewing for each well
Available in full range of colors.
Certified free from Dnase, Rnase, and detectable DNA.