ContactUsLinkComponent
Hareta Replacement Slide Hot Foil Tape, Springside Scientific
Hareta Replacement Slide Hot Foil Tape, Springside Scientific
Catalog # 10756-188
undefined
Hareta Replacement Slide Hot Foil Tape, Springside Scientific
Catalog # 10756-188
Supplier Number:  HSFT300
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Dimensions:
    25 mm x 300 m
  • Cat. No.:
    10756-188
  • Packaging:
    Round Metal Box
  • Supplier no.:
    HSFT300
Specifications
About this item
This tape is used for automated slide printing and designed for use on Thermo SlideMate printers to replace the expensive tape currently offered by the OEM.
Prints up to 12,000 slides per roll. Will not fade, even in the harshest processing environments, including alcohols and xylene. A direct replacement for the original OEM tape.
Packaging: Each roll is 25 mm x 300 m in length and is a long lasting, standard sized 984' roll.