Anti-PFKFB1 Rabbit Polyclonal Antibody
80085-040
: Abgent
- Antibody Type:Primary
- Antigen Name:6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1
- Antigen Symbol:PFKFB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal (1–100 aa)
- Cross Adsorption:No
- Antigen Synonyms:6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1|F6PK|6PF-2-K/Fru-2,6-P2ASE liver isozyme
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80085-040
- Supplier no.:AP8147A
Type: Primary
Antigen: PFKFB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse