Anti-MAP2K7 Rabbit Polyclonal Antibody
80084-534
: Abgent
- Antibody Type:Primary
- Antigen Name:Mitogen-Activated Protein Kinase Kinase 7
- Antigen Symbol:MAP2K7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal (402–419 aa)
- Cross Adsorption:No
- Antigen Synonyms:PRKMK7|MEK 7|SAPKK-4|JNKK2|SAPKK4|MKK7|MAPKK7|MEK
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80084-534
- Supplier no.:AP7918A
Type: Primary
Antigen: MAP2K7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (402–419 aa)
Host: Rabbit
Isotype:
Reactivity: Human