Anti-GCK Rabbit Polyclonal Antibody
80084-496
: Abgent
- Antibody Type:Primary
- Antigen Name:Glucokinase
- Antigen Symbol:GCK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal (1–100 aa)
- Cross Adsorption:No
- Antigen Synonyms:HXKP|LGLK|MODY2|Hexokinase type IV|GK|HK4|HHF3|FGQTL3|Hexokinase D|GLK|HKIV
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80084-496
- Supplier no.:AP7901A
Type: Primary
Antigen: GCK (glucokinase (hexokinase 4))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse