Anti-IRAK3 Rabbit Polyclonal Antibody
80084-446
: Abgent
- Antibody Type:Primary
- Antigen Name:IRAK3 (interleukin-1 Receptor-associated kinase 3)
- Antigen Symbol:IRAK3 (interleukin-1 Receptor-associated kinase 3)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal (60–77 aa)
- Cross Adsorption:No
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80084-446
- Supplier no.:AP7804A
Type: Primary
Antigen: IRAK3 (interleukin-1 receptor-associated kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human