Anti-DOK5 (docking Protein 5) Rabbit Polyclonal Antibody
Catalog # 80084-356
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Symbol:DOK5 (docking Protein 5)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal (291–306 aa)
- Cross Adsorption:No
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80084-356
- Supplier no.:AP7693B
Specifications
About this item
Type: Primary
Antigen: DOK5 (docking Protein 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (291–306 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse