Anti-ILK1/ILK2 Rabbit Polyclonal Antibody
Catalog # 80084-258
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Integrin-Linked Kinase
- Antigen Symbol:ILK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal (26–42 aa)
- Cross Adsorption:No
- Antigen Synonyms:ILK1|Integrin-linked protein kinase|ILK2|p59ILK |59 kDa serine/threonine-protein kinase|ILK-1|ILK-2
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80084-258
- Supplier no.:AP7651A
Specifications
About this item
Type: Primary
Antigen: ILK1/ILK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (26–42 aa)
Host: Rabbit
Isotype:
Reactivity: Human