Anti-ERBB2 Rabbit Polyclonal Antibody
Catalog # 80084-220
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Erb-b2 receptor tyrosine kinase 2
- Antigen Symbol:ERBB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal (1221–1236 aa)
- Cross Adsorption:No
- Antigen Synonyms:NEU|TKR1|MLN 19|CD340|HER2|HER-2/neu|HER-2|NGL
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80084-220
- Supplier no.:AP7629B
Specifications
About this item
Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (1221–1236 aa)
Host: Rabbit
Isotype:
Reactivity: Human