Anti-PRKAA2 Rabbit Polyclonal Antibody
Catalog # 80083-900
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Kinase AMP-activated Catalytic Subunit Alpha 2
- Antigen Symbol:PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal (450–550 aa)
- Cross Adsorption:No
- Antigen Synonyms:AMPKa2|AMPK|AMPK2|PRKAA
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80083-900
- Supplier no.:AP7203A
Specifications
About this item
Type: Primary
Antigen: PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (450–550 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse