Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-MMP26 Rabbit Polyclonal Antibody
Catalog # 80089-058
Supplier:  Abgent
Anti-MMP26 Rabbit Polyclonal Antibody
Catalog # 80089-058
Supplier:  Abgent
Supplier Number:  AP6207A

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Matrix Metalloproteinase 26
  • Antigen Symbol:
    MMP26
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Epitope:
    N-Terminal
  • Cross Adsorption:
    No
  • Antigen Synonyms:
    MGC126590|MGC126592
  • Storage Temperature:
    Cool
  • Shipping Temperature:
    Ice
  • Cat. No.:
    80089-058
  • Supplier no.:
    AP6207A

Specifications

About this item

Type: Primary
Antigen: MMP26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human