Anti-GPLD1 (glycosylphosphatidylinositol specific phospholipase D1) Rabbit Polyclonal Antibody
Catalog # 80088-602
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Glycosylphosphatidylinositol Specific Phospholipase D1
- Antigen Symbol:GPLD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Size:400 µL
- Epitope:N-Terminal ( 1–50 aa)
- Cross Adsorption:No
- Antigen Synonyms:Glycosyl-phosphatidylinositol-specific phospholipase D|PLD|PIGPLD|Glycoprotein phospholipase D|GPIPLDM|GPIPLD|Phosphatidylinositol-glycan-specific phospholipase D|PI-G PLD|PIGPLD1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80088-602
- Supplier no.:AP2405A
Specifications
About this item
Type: Primary
Antigen: GPLD1 (glycosylphosphatidylinositol specific phospholipase D1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human