Anti-USP3 Rabbit Polyclonal Antibody
Catalog # 80083-628
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin specific peptidase 3
- Antigen Symbol:USP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Cross Adsorption:No
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80083-628
- Supplier no.:AP2132C
Specifications
About this item
Type: Primary
Antigen: USP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human