Anti-USP2 Rabbit Polyclonal Antibody
Catalog # 80083-620
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin specific peptidase 2
- Antigen Symbol:USP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal ( 50–150 aa)
- Cross Adsorption:No
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80083-620
- Supplier no.:AP2131A
Specifications
About this item
Type: Primary
Antigen: USP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 50–150 aa)
Host: Rabbit
Isotype:
Reactivity: Human