Anti-UBL4A Rabbit Polyclonal Antibody
Catalog # 80083-614
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin-like 4A
- Antigen Symbol:UBL4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Cross Adsorption:No
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80083-614
- Supplier no.:AP2129B
Specifications
About this item
Type: Primary
Antigen: UBL4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human