Anti-UCHL5 Rabbit Polyclonal Antibody
Catalog # 80083-608
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase isozyme L5
- Antigen Symbol:UCHL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal ( 1–100 aa)
- Cross Adsorption:No
- Antigen Synonyms:(Ubiquitin C-terminal hydrolase L5|Ubiquitin C-terminal hydrolase|CGI-70|UCH-L5|Ubiquitin thioesterase L5|AD-019|UCH37
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80083-608
- Supplier no.:AP2128A
Specifications
About this item
Type: Primary
Antigen: UCHL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse