Anti-Reptin/TIP49B/RUVB2 Rabbit Polyclonal Antibody
Catalog # 60941-430
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Symbol:Reptin/TIP49B/RUVB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:Center
- Cross Adsorption:No
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:60941-430
- Supplier no.:AP1922B
Specifications
About this item
Type: Primary
Antigen: Reptin/TIP49B/RUVB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human