Anti-SETD7 Rabbit Polyclonal Antibody
Catalog # 60941-210
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:SET Domain Containing 7
- Antigen Symbol:SETD7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Cross Adsorption:No
- Antigen Synonyms:histone H3-lysine 4-specific methyltransferase|KIAA1717|lysine N-methyltransferase 7|histone-lysine N-methyltransferase SETD7|KMT7|SET7/9|H3-K4-HMTase SETD7|SET domain-containing protein 7|SET9|SET7|FLJ21193|histone H3-K4 methyltransferase SETD7
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:60941-210
- Supplier no.:AP1194D
Specifications
About this item
Type: Primary
Antigen: SETD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human