Anti-MBD3 Rabbit Polyclonal Antibody
80087-736
: Abgent
- Antibody Type:Primary
- Antigen Name:Methyl-CpG-binding Domain protein 3
- Antigen Symbol:MBD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:100–200 aa
- Cross Adsorption:No
- Antigen Synonyms:methyl-CpG-binding domain protein 3|CpG-binding domain protein 3|CpG-binding domain 3
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80087-736
- Supplier no.:AP1038A
Type: Primary
Antigen: MBD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 100–200 aa
Host: Rabbit
Isotype:
Reactivity: Human