Order Entry
Puerto Rico
ContactUsLinkComponent
Tween® 80 (Polysorbate) 0.1% in deionized water
Tween® 80 (Polysorbate) 0.1% in deionized water
Catalog # 10436-332
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Tween® 80 (Polysorbate) 0.1% in deionized water
Catalog # 10436-332
Supplier:  Hardy Diagnostics
Supplier Number:  U259
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Size:
    125 ml
  • Cat. No.:
    10436-332
  • Supplier no.:
    U259

Specifications

About this item

 :
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:9005-65-6
MDL Number:MFCD00082107