Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody [clone: 5A9F9] (FITC (Fluorescein Isothiocyanate))
Catalog # 89495-094
Supplier: Genscript
Specifications
- Antibody Type:Tag
- Antigen Name:NWSHPQFEK Tag
- Antigen Symbol:NWSHPQFEK Tag
- Clonality:Monoclonal
- Clone:5A9F9
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Size:100 μg
- Cross Adsorption:No
- Form:Lyophilized
- Storage Buffer:PBS, pH 7.4, containing 10 mg/ml BSA and 0.02% sodium azide
- Storage Temperature:The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml
- Shipping Temperature:Ice
- Immunogen:NWSHPQFEK peptide conjugated to KLH
- Purification:Protein A affinity column
- Cat. No.:89495-094
- Packaging:Vial
- Supplier no.:A01736-100
Specifications
About this item
THE™ NWSHPQFEK Tag antibody [FITC], mAb, Mouse recognizes N-terminal and C-terminal Strep II tagged fusion proteins.
- Comprehensive pool
- Against 24 tags
- Available in gram level bulk quantities
- Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
Type: Tag
Antigen: NWSHPQFEK Tag
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: