Anti-V5 Tag Mouse Monoclonal Antibody [clone: 4C12E11]
Catalog # 89495-122
Supplier: Genscript
Specifications
- Antibody Type:Tag
- Antigen Name:V5 Tag
- Antigen Symbol:V5 Tag
- Clonality:Monoclonal
- Clone:4C12E11
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Western Blot:Yes
- Size:100 μg
- Cross Adsorption:No
- Form:Lyophilized
- Storage Buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage Temperature:The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml
- Shipping Temperature:Ice
- Immunogen:V5 tag peptide conjugated to KLH
- Purification:Protein A affinity column
- Cat. No.:89495-122
- Packaging:Vial
- Supplier no.:A01724-100
Specifications
About this item
THE™ V5 Tag antibody, mAb, Mouse recognizes C-terminal, N-terminal and internal V5 tagged fusion proteins.
- Comprehensive pool
- Against 24 tags
- Available in gram level bulk quantities
- Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
Type: Tag
Antigen: V5 Tag
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: