Anti-Streptavidin Rabbit Polyclonal Antibody
Catalog # 89494-990
Supplier: Genscript
Specifications
- Antibody Type:Primary
- Antigen Name:Streptavidin
- Antigen Symbol:Streptavidin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:5 mg
- Cross Adsorption:No
- Form:Lyophilized
- Storage Buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage Temperature:The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 5 mg/ml
- Shipping Temperature:Ice
- Immunogen:Purified recombinant streptavidin
- Tested Applications:Dot blot
- Purification:Protein A/G affinity column
- Cat. No.:89494-990
- Packaging:Vial
- Supplier no.:A00621-5
Specifications
About this item
GenScript Streptavidin Antibody is specific to streptavidin.
- High specificit
- High affinity
- High stability
GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.
Type: Primary
Antigen: Streptavidin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: