About this item
Criterion™ IEF Precast Gels are available in broad range (pH 3–10) or narrow range (pH 5–8) with different numbers of wells and well volumes
- pH 3–10 optimal separation range: ~pH 3.5–8.5
- pH 5–8 optimal separation range: ~pH 4.6–7.8
Separate soluble native proteins in only 150 minutes when using a programmable high-voltage power supply.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 101645-856
- 101645-860
- 101645-854
- Buffer Volume:
- Upper, 60 mL; Lower, 400 mL
- Upper, 60 mL; Lower, 400 mL
- Upper, 60 mL; Lower, 400 mL
- Cassette Dimensions:
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- Cassette Material:
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Comb Material:
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Format:
- Midi, IEF
- Midi, IEF
- Midi, IEF
- Gel Dimensions:
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- Resolving Gel:
- 5% (no stacker)
- 5% (no stacker)
- 5% (no stacker)
- Shelf Life:
- 26 weeks
- 26 weeks
- 26 weeks
- Storage Conditions:
- 4°C store flat; do not freeze
- 4°C store flat; do not freeze
- 4°C store flat; do not freeze
- Tray Material:
- PET
- PET
- PET