Criterion™ XT Bis-Tris Precast Gels are formulated using a Bis-Tris buffer system (pH 6.4) and can be run with either XT MOPS or XT MES running buffers
- Useful for screening new samples and evaluating sample preparation conditions
- Laemmli-type separation
- Run 2-D gels in less than a day
- Near-neutral pH delays acrylamide hydrolysis significantly longer than Laemmli buffers
- Very stable, with a 12 month shelf life at room temperature
The ability to use two different buffer systems provides the flexibility to obtain different separation profiles with only one type of gel. XT MOPS buffer system is commonly used for mid-sized proteins and XT MES buffer system for smaller proteins.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 101645-928
- 101645-922
- 101645-918
- 101645-908
- 101645-926
- 101645-906
- 101645-924
- 101645-914
- 101645-904
- 101645-920
- 101645-902
- 101645-912
- 101645-910
- Running Buffer:
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- XT MOPS or XT MES
- Sample Buffer:
- XT
- XT
- XT
- XT
- XT
- XT
- XT
- XT
- XT
- XT
- XT
- XT
- XT
- Cassette Dimensions:
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- Cassette Material:
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Comb Material:
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Format:
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Gel Dimensions:
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- Shelf Life:
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- Storage Conditions:
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Store flat at room temperature; do not freeze
- Tray Material:
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET
- PET