Floor Mat Ground Cord, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
The floor mat ground cord series of low profile ground cords are highly durable and reliable ground cords
- 15’ cord length
- 10 mm (3/8”) male snap or 10 mm (3/8”) female snap
- #10 ring type ground terminal
- PU/PVC wire insulation
- 1 meg ohm resistor
- Black cord
A molded low profile mushroom cap ensures secure connection to floor mats and easily attaches to floor mats. Its molded dome cap reduces tripping and allows chairs to easily roll over them without getting snagged. The low profile, molded head has built in snap - your choice of male or female - for a secure work surface attachment. A 15' black ground cord terminate with a standard 1/8" (#10) ring terminal for a solid connection to ground. Also available without a resistor.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20.