ContactUsLinkComponent
GeneMate Combination Packs, 12-Strip Standard Tubes with 12-Strip Optically-Clear Flat Caps, Scientific Specialties
GeneMate Combination Packs, 12-Strip Standard Tubes with 12-Strip Optically-Clear Flat Caps, Scientific Specialties
Catalog # 490003-628
Supplier:  GeneMate
undefined
GeneMate Combination Packs, 12-Strip Standard Tubes with 12-Strip Optically-Clear Flat Caps, Scientific Specialties
Catalog # 490003-628
Supplier:  GeneMate
Specifications
  • Description:
    12-Strip Tubes and Caps
  • Cat. No.:
    490003-628
  • Supplier no.:
    3136-00
Specifications
About this item
Designed to precisely fit most thermal cyclers. Ideal for qPCR (real-time PCR). Convenient tube and cap combination packs. Equal number of tubes and caps in one pack.
  • Secure evaporation-resistant seal between cap lid and tube
  • Optically-clear flat caps
  • Fluorescent signal is passed through unaffected
  • Ultra-thin wall for optimum heat transfer and higher yield
  • Tubes are pre-numbered for easy well identification
  • Crystal-clear clarity, easy to visualize content of each well
  • Available in white
Ideal for enhancing signal strength for qPCR assays.
Certified free DNase, RNase, and detectable DNA.
Ordering information: GeneMate 96-well PCR tube storage racks are available separately.