HLT™ ERGO-F, Pipette Tip, Thermo Scientific
Catalog # 72830-666
Supplier: Thermo Scientific
Specifications
- Volume:200 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental Certification:ISO14001 (Certificate Registration No. EMS 752603)
- Cat. No.:72830-666
- Tip style:HLT™ 200 Ergo-F, clear
- For use with:Finnpipette®
- Supplier no.:3812