About this item
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Stand up cup that detects drug metabolites in urine with results in 5 minutes. Cup and temperature strip included, SVT option available.
Room temperature storage, long shelf-life.
Rapid screen for a collection of illicit and prescription drugs. Each kit contains materials for 25 drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Cocaine (COC), Methamphetamine (mAMP), Opiates (OPI) and (MOP), Methadone (MTD), Oxycodone (OXY), Phencyclidine (PCP), Propoxyphene (PPX), Ecstacy (MDMA), Tricyclic Antidepressents (TCA), and Marijuana (THC) in human urine at the following cutoff levels (ng/mL) unless otherwise noted in description: AMP (1000), BAR (300), BZO (300), BUP (10), COC (300), mAMP (1000), OPI (2000), MOP (300), MTD (300), OXY (100), PCP (25), PPX (300), MDMA (500), TCA (1000), and THC (50).