Anti-UCHL1 Mouse Monoclonal Antibody [clone: 1C9E11]
Catalog # 10753-376
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen Symbol:UCHL1
- Clonality:Monoclonal
- Clone:1C9E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Pig,Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Cross Adsorption:No
- Antigen Synonyms:Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Recombinant Protein
- Purification:Protein G purification
- Cat. No.:10753-376
- Supplier no.:66230-1-IG
Specifications
About this item
The UCHL1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human UCHL1. This antibody recognizes human, mouse, pig antigen. The UCHL1 antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.
Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: 1C9E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Pig