Anti-CD13 Mouse Monoclonal Antibody [clone: 2D8D11]
Catalog # 10221-848
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Alanyl Aminopeptidase
- Antigen Symbol:ANPEP
- Clonality:Monoclonal
- Clone:2D8D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Cross Adsorption:No
- Antigen Synonyms:Apn|AP-N|Cd13|aminopeptidase M|membrane protein p161|aminopeptidase N|microsomal aminopeptidase|aminopeptidase N/CD13|alanyl aminopeptidase|P150|AP-M
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Recombinant Protein
- Purification:Protein A purification
- Cat. No.:10221-848
- Supplier no.:66211-1-IG
Specifications
About this item
The CD13 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human CD13. This antibody recognizes human antigen. The CD13 antibody has been validated for the following applications: ELISA, WB, IHC analysis.
Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: 2D8D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human