To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Suppression of tumorigenicity 6
- Antigen Symbol:ST6
- Clonality:Monoclonal
- Clone:1A7B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Recombinant Protein
- Purification:Protein G purification
- Cat. No.:10351-938
- Supplier no.:66209-1-IG
The ST6GALNAC6 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human ST6GALNAC6. This antibody recognizes human antigen. The ST6GALNAC6 antibody has been validated for the following applications: ELISA, WB analysis.
Type: Primary
Antigen: ST6 (alpha-N-acetyl-neuraminyl-2 3-beta-galactosyl-1 3)-N-acetylgalactosaminide alpha-2 6-sialyltransferase 6
Clonality: Monoclonal
Clone: 1A7B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human