Anti-FOXC1 Rabbit Polyclonal Antibody
10220-778
- Antibody Type:Primary
- Antigen Name:Forkhead Box C1
- Antigen Symbol:FOXC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Cross Adsorption:No
- Antigen Synonyms:IHG1|FKHL7|IGDA|IRID1|FREAC-3|FREAC3|RIEG3|ARA
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Peptide
- Purification:Antigen affinity purification
- Cat. No.:10220-778
- Supplier no.:55365-1-AP
The FOXC1 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human FOXC1. This antibody recognizes human, mouse antigen. The FOXC1 antibody has been validated for the following applications: ELISA, WB analysis.
Type: Primary
Antigen: FOXC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse