Anti-CD68 Rabbit Polyclonal Antibody
Catalog # 10220-880
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:CD68 molecule
- Antigen Symbol:CD68
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:150 µL
- Cross Adsorption:No
- Antigen Synonyms:macrosialin|CD68 antigen|GP110|scavenger receptor class D member 1|SCARD1|LAMP4|macrophage antigen CD68
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Fusion Protein
- Purification:Antigen affinity purification
- Cat. No.:10220-880
- Supplier no.:25747-1-AP
Specifications
About this item
The CD68 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CD68. This antibody recognizes human antigen. The CD68 antibody has been validated for the following applications: ELISA, IHC analysis.
Type: Primary
Antigen: CD68
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: