Anti-TACC3 Rabbit Polyclonal Antibody
Catalog # 10222-106
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Transforming acidic coiled-coil-Containing protein 3
- Antigen Symbol:TACC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Cross Adsorption:No
- Antigen Synonyms:Transforming acidic coiled-coil-containing protein 3|TACC3|ERIC-1|ERIC1
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Fusion Protein
- Purification:Antigen affinity purification
- Cat. No.:10222-106
- Supplier no.:25697-1-AP
Specifications
About this item
The TACC3 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human TACC3. This antibody recognizes human antigen. The TACC3 antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.
Type: Primary
Antigen: TACC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human