Anti-SLCO4C1 Rabbit Polyclonal Antibody
10220-924
- Antibody Type:Primary
- Antigen Name:Solute Carrier Organic Anion Transporter Family member 4C1
- Antigen Symbol:SLCO4C1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Cross Adsorption:No
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Fusion Protein
- Purification:Antigen affinity purification
- Cat. No.:10220-924
- Supplier no.:24584-1-AP
The SLCO4C1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SLCO4C1. This antibody recognizes human, mouse antigen. The SLCO4C1 antibody has been validated for the following applications: ELISA, WB analysis.
Type: Primary
Antigen: SLCO4C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse