Anti-BARD1 Rabbit Polyclonal Antibody
Catalog # 10273-124
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:BRCA1-associated RING Domain protein 1
- Antigen Symbol:BARD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Cross Adsorption:No
- Antigen Synonyms:BRCA1-associated RING domain protein 1|BARD-1|BARD1
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Fusion Protein
- Purification:Antigen Affinity purified
- Cat. No.:10273-124
- Supplier no.:22964-1-AP
Specifications
About this item
The BARD1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human BARD1. This antibody recognizes human antigen. The BARD1 antibody has been validated for the following applications: ELISA, WB analysis.
Type: Primary
Antigen: BARD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human