Anti-CDX1 Rabbit Polyclonal Antibody
Catalog # 10220-854
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Caudal Type Homeobox 1
- Antigen Symbol:CDX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Cross Adsorption:No
- Antigen Synonyms:Homeobox protein CDX-1|Caudal-type homeobox protein 1
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Fusion Protein
- Purification:Antigen affinity purification
- Cat. No.:10220-854
- Supplier no.:21655-1-AP
Specifications
About this item
The CDX1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CDX1. This antibody recognizes human, mouse antigen. The CDX1 antibody has been validated for the following applications: ELISA, WB analysis.
Type: Primary
Antigen: CDX1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: