Anti-DNAJB7 Rabbit Polyclonal Antibody
Catalog # 10220-910
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:DnaJ (Hsp40) Homolog, Subfamily B, Member 7
- Antigen Symbol:DNAJB7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Cross Adsorption:No
- Antigen Synonyms:DJ5|HSC3
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Fusion Protein
- Purification:Antigen affinity purification
- Cat. No.:10220-910
- Supplier no.:18540-1-AP
Specifications
About this item
The DNAJB7 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human DNAJB7. This antibody recognizes human,mouse,rat antigen. The DNAJB7 antibody has been validated for the following applications: ELISA, WB analysis.
Type: Primary
Antigen: DNAJB7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat