Anti-MFSD10 Rabbit Polyclonal Antibody
10272-978
- Antibody Type:Primary
- Antigen Name:Major Facilitator Superfamily Domain Containing 10
- Antigen Symbol:MFSD10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Cross Adsorption:No
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity purification
- Cat. No.:10272-978
- Supplier no.:11518-1-AP
The MFSD10 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human MFSD10. This antibody recognizes human,mouse,rat antigen. The MFSD10 antibody has been validated for the following applications: ELISA, WB analysis.
Type: Primary
Antigen: MFSD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat