About this item
Criterion™ TGX Stain-Free™ Precast Gels for PAGE are based on the long shelf life TGX (Tris-Glycine eXtended) formulation and allow rapid fluorescent detection of proteins, after both electrophoresis and blotting, with stain-free enabled imaging systems (Bio-Rad’s Gel Doc™ EZ, Gel Doc™ XR+, ChemiDoc™ Touch, ChemiDoc™ MP, and ChemiDoc™ XRS+ Imaging Systems).
- Fast run times
- Protein separation, gel imaging, and analysis in less than 30 minutes
- Gel images and complete analysis in less than 5 minutes after electrophoresis
- Verify transfer to membranes with a stain-free enabled imager
- Use standard sample and running buffers for Laemmli-like separations
- Comparable sensitivity to Coomassie stain
- Better reproducibility and quantitation compared to other staining procedures
- Same gel can be used for staining, western blotting, or mass spectrometry
TGX Stain-Free™ Gels retain Laemmli-like separation characteristics when using standard sample and Tris/glycine running buffers. Proteins can be separated in as little as 20 minutes, and then visualized in 2.5 minutes. Within gels or on PVDF membranes, trihalo compounds react with tryptophan residues in a UV-induced reaction to produce fluorescence, which can be easily detected by the stain-free enabled imagers.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 101764-862
- 101764-860
- 101764-858
- 101764-856
- 101646-120
- 101764-872
- 101764-870
- 101764-868
- 101764-866
- 101764-864
- 101646-118
- 101646-102
- 101646-098
- 101646-116
- 102650-412
- 102650-410
- 101646-104
- 101646-096
- 101646-114
- 101764-878
- 101646-108
- 101764-876
- 101646-100
- 101764-874
- 101646-106
- 101646-094
- 101646-092
- 101646-110
- 101646-112
- Running Buffer:
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- Sample Buffer:
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- Cassette Dimensions:
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- Cassette Material:
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Comb Material:
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Format:
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Gel Dimensions:
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- Gel Type:
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- TGX (Tris-glycine eXtended) Stain-Free™
- Run Time:
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- Shelf Life:
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- Storage Conditions:
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze