About this item
Criterion™ TGX™ (Tris-Glycine eXtended) Precast Gels for PAGE are based on the long shelf life TGX formulation
- Run time as short as 20 min
- Use standard sample and running buffers for Laemmli-like separations
- High-throughput system
TGX™ Gels are manufactured without SDS. For standard denaturing separations, gels are run with SDS containing running buffer, typically Tris/glycine/SDS. For nondenaturing conditions, SDS is omitted from the running buffer.
TGX™ Gels retain Laemmli-like separation characteristics using standard sample and Tris/glycine running buffers. Proteins can be separated in as little as 20 min at 300 V while maintaining low temperatures.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 101693-672
- 101693-682
- 101764-854
- 101693-668
- 101764-852
- 101693-674
- 101693-680
- 101693-690
- 101693-666
- 102650-376
- 101693-686
- 101764-838
- 101764-836
- 101764-834
- 101764-832
- 101693-670
- 101693-676
- 101693-678
- 101693-664
- 101693-662
- 101764-850
- 101764-848
- 101764-846
- 101764-844
- 101764-842
- 101764-840
- 101693-684
- 101693-688
- 102650-388
- 102650-386
- Running Buffer:
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- 25 mM Tris, 192 mM glycine, 0.1% SDS, pH 8.3
- Sample Buffer:
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- 62.5 mM Tris-HCl, pH 6.8, 2% SDS, 25% glycerol, 0.01% bromophenol blue
- Cassette Dimensions:
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- 15.0 x 10.6 x 0.53 mm
- Cassette Material:
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Styrene copolymer
- Comb Material:
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Format:
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Midi
- Gel Dimensions:
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- 13.3 x 8.7 x 0.1 mm
- Gel Type:
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- TGX (Tris-glycine eXtended)
- Run Time:
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- 42–50 min (at 200V constant); 20–26 min (at 300V constant)
- Shelf Life:
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- 12 Months
- Storage Conditions:
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze
- Store flat at 4°C; do not freeze