11 mm Wide Opening Plastic Crimp/Snap Top Vials, Thermo Scientific
Supplier: Avantor
Certificates
About this item
Choose from these chemically inert polypropylene vials, manufactured from high-quality, GC-tested polypropylene material, that are suitable for most chromatography applications.
11 mm Plastic Crimp/Snap Top Vials provide an economical alternative to glass for some applications. Polypropylene is chemically inert and suitable for most chromatography applications, and can be used when levels of inorganic ions have to be kept to an absolute minimum.