Order Entry
Puerto Rico
ContactUsLinkComponent
Shelf, PVC, Benchtop with Safety Lip, TrippNT
Shelf, PVC, Benchtop with Safety Lip, TrippNT
Catalog # 10012-620
Supplier:  TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Shelf, PVC, Benchtop with Safety Lip, TrippNT
Catalog # 10012-620
Supplier:  TrippNT
Supplier Number:  50113
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Depth:
    6 ¾"
  • Width:
    24"
  • Description:
    Straight, Triple
  • Height:
    16"
  • Cat. No.:
    10012-620
  • Weight:
    7.2 lbs.
  • Supplier no.:
    50113

Specifications

About this item

The workhorse of the lab TrippNT white PVC safety shelves create more space, higher order and nicer overall appearance. With no assembly required and a lifetime full replacement guarantee, TrippNT lab shelves organize, sort and separate expensive reagents, chemicals, supplies and tools. With over 100 shelves sizes there is one to fit every need.

  • Variety of shelves to keep items organized, safe, clean
  • Use on bench, counter, cabinet, or floor
  • Several sizes and designs
  • White PVC construction
  • Safety lip to keep items safe
  • Durable and easy to clean
  • In stock and ready to ship
  • American made by TrippNT

Angled or straight sides accommodate space and sorting options.